missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers

Individual Calibration Report as needed for many standard operating procedures.

Supplier:  H-B Instrument™ B602020100

Catalog No. S02698


Only null left
Explore available promotions

Description

Description

  • Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.
  • Individually serialized
  • Made in the USA
Specifications

Specifications

Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA
Partial Immersion
12 in.
Glass
110°C
-20°C to +110°C
1°C
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Detailed calibration report
300 mm
Organic
-20°C
±1°C
76 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions