Learn More
H-B Instrument™ Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion

Individual Calibration Report as needed for many standard operating procedures
Supplier: H-B Instrument™ B602020000
Description
- Fill: Black spirit
- Each thermometer is individually calibrated
- Celsius-scale
- Designed to be very easy to read
- Replace mercury thermometers with the same accuracy without any hazards
- Nontoxic, nonhazardous, biodegradable
- NIST-Certified detailed, laminated report of calibration
- Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read
- Individually calibrated to NIST standards at specific temperature and supplied with NIST report of calibration stating corrections to one-tenth the smallest division
- Contain an environmentally friendly non-mercury liquid that is non-toxic, non-hazardous, and fully biodegradable
- In case of breakage the liquid can be cleaned up easily with soap and water
- “Lead-free”, yellow back glass and black liquid provides easy readability and fewer reading errors
- Contains non-toxic, biodegradable, Enviro-Safe™ liquid
- EnviroKleen™ certified
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Certifications
Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST™ standards used for the calibration. Individually serialized and made in the USA.
Chemical Identifiers
| Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA | |
| Precision | |
| Glass | |
| 76 mm |
| Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. | |
| Partial Immersion | |
| Enviro-Safe™ Liquid |
Specifications
| 0, 37, 56°C | |
| Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. | |
| Partial Immersion | |
| 300 mm | |
| Enviro-Safe™ Liquid | |
| -10°C | |
| ±0.5°C | |
| 76 mm |
| Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA | |
| Precision | |
| 12 in. | |
| Glass | |
| 70°C | |
| -10°C to +70°C | |
| 0.5°C |