Learn More
H-B Instrument™ Durac™ Plus™ Precision Verification Thermometers
To verify sample temperatures in freezers, refrigerators, incubators and ovens.
$110.00 - $286.14
Specifications
Bottle Fill Glass Beads
Certifications/Compliance ISO/IEC 17025:2005, A2LA, NIST, DAkkS
Includes Multilanguage instructions, statement of accuracy indicating accuracy traceable to NIST
Thermometer Fill Blue Organic Liquid
Product Type Precision Verification Thermometer
View More Specs
Includes: Multilanguage (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Products 8
Catalog Number Mfr. No. Temperature Range (Metric) Bottle Size For Use With (Application) Length (Metric) Overall Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Bottle Size For Use With (Application) Length (Metric) Overall Price Quantity  
13201565
View Documents
H-B Instrument™
B606000500RMD
-30°C to +1°C 30 mL Freezers 135 mm
Each for $111.00
Only null left
13201566
View Documents
H-B Instrument™
B606000600RMD
-5°C to +15°C 60 mL Refrigerator 125 mm
Each for $117.13
Only null left
13201567
View Documents
H-B Instrument™
B606001000RMD
15°C to 50°C 30 mL Incubator 135 mm
Each for $110.00
Only null left
13201584
View Documents
H-B Instrument™
B606000200
15°C to 30°C 60 mL Incubator 180 mm
Each for $187.91
Only null left
13-201-583
View Documents
N/A -2°C to +10°C 60 mL Refrigerator 180 mm
Each for $199.93
Only null left
13201582
View Documents
H-B Instrument™
B606000000
-25°C to +5°C 60 mL Freezers 210 mm
Each for $190.78
Only null left
13201585
View Documents
H-B Instrument™
B606000300
95°C to 115°C 60 mL Oven 200 mm
Each for $230.59
Only null left
13-201-586
View Documents
N/A 25°C to 45°C 60 mL Incubator 190 mm
Each for $286.14
Only null left
Description
Description
Blue liquid fill on white backed glass makes it easier to confirm temperature in freezers, refrigerators, incubators.
Applications:
Ideal for chemical industry, petrochemistry, pharmaceutical, medical, cosmetic, producers of chemicals, university, dairies, breweries, distillers, beer cellars, refrigeration house, hot houses, scientific institutes, blood banks, refrigeration/freezer/incubator/oven equipment producers and producers of
laboratory equipment.
Warranty
Five-year limited. See hbinstrument.com/conditions.php for details.
Specifications
Specifications
Glass Beads
Multilanguage instructions, statement of accuracy indicating accuracy traceable to NIST
Precision Verification Thermometer
ISO/IEC 17025:2005, A2LA, NIST, DAkkS
Blue Organic Liquid
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'