Learn More
H-B Instrument™ Durac™ Plus™ ASTM-Like Partial Immersion Laboratory Thermometers
Safer than their mercury counterparts
Description
- Mercury free, these ASTM-Like thermometers are safer to use than their mercury counterparts
- Ideal for use in quality control laboratories or when mercury filled thermometers cannot be used, but ASTM standards are still needed
- Feature blue organic liquid fill against lead-free white back glass
Chemical Identifiers
| ASTM | |
| Quality control laboratories or where ISO 9000 standards are in place | |
| Glass | |
| Blue Organic Liquid |
Specifications
| 91C | |
| Quality control laboratories or where ISO 9000 standards are in place | |
| 14.6 in. | |
| Glass | |
| 50°C | |
| 20°C to 50°C | |
| 0.1°C | |
| Celsius |
| ASTM | |
| Partial Immersion | |
| 370 mm | |
| Blue Organic Liquid | |
| 20°C | |
| 0.3°C | |
| 76 mm |