missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Durac™ Maximum-Registering Autoclavable Bimetal Thermometers

Indicate the highest temperature achieved and hold that measurement until manually reset

 View more versions of this product

Catalog No. 13-202-761

Only null left
Explore available promotions

Includes: Maximum-registering thermometer and multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy and instructions. 13-202-760 includes an individual Thermometer Calibration Report.

Description

Description

A maximum-registering pointer (red) is moved up by the temperature-measuring pointer (black) to indicate the highest temperature achieved.


  • Threaded connection (1/4 in. NPT) screws directly into in-line processes or use to mount thermometer on included stand
  • Waterproof and pressurized
  • Withstands autoclave cycles
  • Constructed from durable 304 stainless steel with a glass lens
  • Withstands pressure, high temperatures and rust
  • Probe length: 50mm (2 in.)
  • Includes multilanguage (English, French, German, Italian, Portuguese and Spanish) instructions and Statement of Test and Calibration traceable to NIST™

Compliance

Traceable to NIST

Specifications

Specifications

Traceable to NIST
81 mm
2 in.
Glass
300°F
0°F
0°F to 300°F
+/-1.5°C (+/-3°F) full range
3.19 in.
No Calibration Report
50 mm
Bimetal
150°C
-20°C
-20°C to +150°C
2°C/5°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions