missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Double-Safe™ PFA Safety Coated, Liquid-In-Glass Thermometers: Partial Immersion

Protect your workplace and ensure a safe environment

$104.52 - $162.03

Specifications

Certifications/Compliance Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Grade Precision
Immersion Type Partial Immersion
Includes Multilanguage instructions, statement of accuracy
View More Specs

Products 6
Catalog Number Mfr. No. Temperature Range (Metric) Temperature Range (English) Length (Metric) Probe Immersion Length Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Temperature Range (English) Length (Metric) Probe Immersion Length Price Quantity  
13201472
View Documents
H-B Instrument™
B604000800
-10°C to +260°C n.a. 350 mm 76 mm
Each for $162.03
Only null left
 
13201471
View Documents
H-B Instrument™
B604000500
-10°C to +70°C n.a. 300 mm 76 mm
Each for $151.28
Only null left
 
13-202-513
View Documents
N/A -10°C to +150°C n.a. 200 mm 50 mm (2 in.)
Each for $114.43
Only null left
 
13-202-518
View Documents
N/A n.a. 0°F to 230°F 300 mm 76 mm
Each for $107.11
Only null left
 
13-202-516
View Documents
N/A -20°C to +110°C n.a. 300 mm 76 mm
Each for $108.04
Only null left
 
13-202-517
View Documents
N/A -20°C to +150°C n.a. 300 mm 76 mm
Each for $104.52
Only null left
 
Description

Description

Protect your workplace and ensure a safe environment

  • Environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • Manufactured and tested by our USA quality team

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Traceable to NIST™. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Specifications

Specifications

Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Precision
Multilanguage instructions, statement of accuracy
Enviro-Safe™ Liquid
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial Immersion
Glass
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'