Learn More
SP Bel-Art™ H-B DURAC Calibrated Thermocouple Thermometers
H-B DURAC Calibrated Thermocouple Thermometers; -200/1370°C (-328/2498°F)
 View more versions of this product
Catalog No. 01257125
Only null left
Description
Description
  • Used for temperature measurement in life science research, kilns, gas turbine exhaust, diesel engines, and other industrial processes, H-B Calibrated Thermocouple Thermometers provide measurement throughout a broad temperature range and the ability to utilize multiple probes at one time.
  • Wide temperature range allows accurate monitoring at extreme temperatures
  • Max/min/average temperature recording capability
  • Data hold
  • Includes two K-type beaded wire probes (-20/250°C capability); one is calibrated and serialized
  • Software CD (where included) for real time data collection and logging via USB
  • Bluetooth™ app available for use with B60911-0200
  • Probes for wider temperature ranges available (B60901-1200 & B60901-1300) or use alternatively purchased probes; see chart for probe types accepted by each unit 
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • Individually serialized, CE-marked Note: calibration applies to included probe and tested port only
Specifications
Chemical Identifiers
0°C, 100°C
7.25 in.
-328°F to +2498°F
1.25 in.
18.5 cm
-200°C to +1370°C
Specifications
0°C, 100°C
3 cm
18.5 cm
0.1°C between -200/200°C, otherwise 1°C
-328°F to +2498°F
9 V
6.5 cm
1.25 in.
7.25 in.
4 Probe Inputs/ Displays, RS232 Connection
2.5 in.
-200°C to +1370°C
±(0.2% +1°C) between -200/200°C and 400/1370°C; otherwise ±(0.5% +1°C)
Product Suggestions
Product Suggestions