missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-7 dihydrochloride, Thermo Scientific Chemicals
$337.18 - $1320.45
Chemical Identifiers
| CAS | 108930-17-2 |
|---|---|
| Molecular Formula | C14H19Cl2N3O2S |
| Molecular Weight (g/mol) | 364.285 |
| MDL Number | MFCD00036961 |
| InChI Key | OARGPFMFRLLKPF-UHFFFAOYSA-N |
| Synonym | h-7 dihydrochloride, h-7, dihydrochloride, 1-5-isoquinolinylsulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine, 2hcl, 1-5-isoquinoline-sulfonyl-2-methylpiperazine, 5-2-methylpiperazin-1-ylsulfonyl isoquinoline dihydrochloride, isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride, 1-5-isoquinolylsulfonyl-2-methylpiperazine, chloride, chloride, 1-5-isoquinolinesulfonyl-2-methyl-piperazine dihydrochloride |
| PubChem CID | 73332 |
| ChEBI | CHEBI:82739 |
| IUPAC Name | 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride |
| SMILES | CC1CNCCN1S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61697MA
|
Thermo Scientific Chemicals
J61697MA |
10 mg |
Each for $337.18
|
|
|||||
|
AAJ61697MF
|
Thermo Scientific Chemicals
J61697MF |
50 mg |
Each for $1,320.45
|
|
|||||
Description
Protein kinase inhibitor. Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Protein kinase inhibitor
- Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C
Chemical Identifiers
| 108930-17-2 | |
| 364.285 | |
| OARGPFMFRLLKPF-UHFFFAOYSA-N | |
| 73332 | |
| 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride |
| C14H19Cl2N3O2S | |
| MFCD00036961 | |
| h-7 dihydrochloride, h-7, dihydrochloride, 1-5-isoquinolinylsulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine, 2hcl, 1-5-isoquinoline-sulfonyl-2-methylpiperazine, 5-2-methylpiperazin-1-ylsulfonyl isoquinoline dihydrochloride, isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride, 1-5-isoquinolylsulfonyl-2-methylpiperazine, chloride, chloride, 1-5-isoquinolinesulfonyl-2-methyl-piperazine dihydrochloride | |
| CHEBI:82739 | |
| CC1CNCCN1S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl |
Specifications
| 108930-17-2 | |
| White | |
| MFCD00036961 | |
| 5840763 | |
| OARGPFMFRLLKPF-UHFFFAOYSA-N | |
| 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride | |
| 73332 | |
| 364.29 | |
| H-7 dihydrochloride |
| 191°C to 193°C | |
| C14H19Cl2N3O2S | |
| 10 mg | |
| h-7 dihydrochloride, h-7, dihydrochloride, 1-5-isoquinolinylsulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine, 2hcl, 1-5-isoquinoline-sulfonyl-2-methylpiperazine, 5-2-methylpiperazin-1-ylsulfonyl isoquinoline dihydrochloride, isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride, 1-5-isoquinolylsulfonyl-2-methylpiperazine, chloride, chloride, 1-5-isoquinolinesulfonyl-2-methyl-piperazine dihydrochloride | |
| CC1CNCCN1S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl | |
| 364.285 | |
| CHEBI:82739 | |
| Powder |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only