missing translation for 'onlineSavingsMsg'
Learn More

H-7 dihydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ61697MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61697MF 50 mg
AAJ61697MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61697MF Supplier Thermo Scientific Chemicals Supplier No. J61697MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108930-17-2 | Molecular Formula: C14H19Cl2N3O2S | Molecular Weight: 364.285 g/mol

Protein kinase inhibitor. Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Protein kinase inhibitor
  • Inhibits cyclic nucleotide dependent protein kinase (PKA) and protein kinase C

Chemical Identifiers

CAS 108930-17-2
Molecular Formula C14H19Cl2N3O2S
Molecular Weight (g/mol) 364.285
MDL Number MFCD00036961
InChI Key OARGPFMFRLLKPF-UHFFFAOYSA-N
Synonym h-7 dihydrochloride, h-7, dihydrochloride, 1-5-isoquinolinylsulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine dihydrochloride, 1-5-isoquinolinesulfonyl-2-methylpiperazine, 2hcl, 1-5-isoquinoline-sulfonyl-2-methylpiperazine, 5-2-methylpiperazin-1-ylsulfonyl isoquinoline dihydrochloride, isoquinoline-5-sulfonic 2-methyl-1-piperazide dihydrochloride, 1-5-isoquinolylsulfonyl-2-methylpiperazine, chloride, chloride, 1-5-isoquinolinesulfonyl-2-methyl-piperazine dihydrochloride
PubChem CID 73332
ChEBI CHEBI:82739
IUPAC Name 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride
SMILES CC1CNCCN1S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl

Specifications

Melting Point 191°C to 193°C
Color White
Quantity 50 mg
Beilstein 5840763
Formula Weight 364.29
Physical Form Powder
Chemical Name or Material H-7 dihydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only