Learn More
Supertekā„¢ Gyroscope DFS Item
GREENER_CHOICE
Gyroscope with Gimbal Cradle is an excellent, inexpensive apparatus, which is used to explore the mysteries of precession, inertial guidance, gyroscopic compasses, and stabilization.
Supplier:  Supertekā„¢ PH10589H
Catalog No. S012336
Only null left
Explore available promotions
Description
Description
  • Gyroscope with Gimbal Cradle is an excellent, inexpensive apparatus.
  • Used to explore the mysteries of precession, inertial guidance, gyroscopic compasses, and stabilization.
  • It is precision machined and balanced and runs smoothly with little vibration.
  • Provided with pull string with handle, weight set, and support hook.
  • A pendulum mounting is also supplied that can be used for investigating the rolling moment.
  • It demonstrates the principle of angular momentum.
  • Instructional manual included.
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
    • Non-Toxic
    • Ozone-Safe or Safe for the Ozone Layer
Specifications
Specifications
Gyroscope
Product Suggestions
Product Suggestions