Learn More
Coast GX30 Waterproof LED Flashlight
Catalog No. 17018134
Encompass
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
17018134 LED
1 options
Catalog No. 17018134 Supplier Coast Supplier No. 30938*
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
When you choose Industrial Performance batteries, you get the quality guarantee of a company that’s been at it for 100 years, with products that make people’s work safer and lives easier.
  • Go boldly into the dark, no matter the storm ahead. The GX30 matches massive lighting power with a hard-wearing waterproof build fit for any job or adventure.
  • The ultra-strong, lightweight polymer handle is sealed with tight gaskets to achieve IPX7 waterproof protection down to 1 meter. And four output modes—medium, high, Turbo, and low—peak at an impressive 2300 lumens.
  • The COAST PURE BEAM™ optic and TWIST™ focus system enable seamless transitions between the edge-to-edge clarity of the Ultra View Flood Beam and the long-range precision of the BULLS-EYE™ Spot Beam.
  • The flashlight's grip-textured handle includes a lanyard loop for easy carry or storage, which doubles as an anti-roll tail cap so that you can set the flashlight down and not worry about it getting away on uneven surfaces.
  • Engineered with COAST Dual Power technology, this versatile flashlight runs on six AA alkaline batteries but can also operate using a COAST ZITHION-X™ ZX866 rechargeable battery (purchased separately).
  • The GX30 is constructed of top-grade materials to meet IP67 waterproof and dust-resistant standards and is backed by a lifetime warranty.
Specifications
Light Source LED
Voltage 1.5 V
Color Black
Product Type Streamlight Battery