Learn More
3M™ GVP-Series Belt-Mounted Powered Air-Purifying Respirator (PAPR) Assemblies
GVP-1 PAPR Assembly offers respiratory protection and allows freedom of movement while being used in applications. Includes all the components needed to make a complete 3M PAPR System.
$1,325.91 - $1,605.01
Specifications
Battery Life 8 hours
Filter Type High Efficiency (HE)
Product Type PAPR Assembly, No Headtop
For Use With (Equipment) PAPR respirator
Includes Motor blower, battery, charger, flow meter, power cord, blower plugs
View More Specs
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Products 3
Catalog Number Mfr. No. Belt Material For Use With (Application) Price Quantity  
Catalog Number Mfr. No. Belt Material For Use With (Application) Price Quantity  
19-087398
View Documents
3M™
GVP1
Web Belt Assembly, facility maintenance, machine operation, painting, sanding N/A
Only null left
29-635-0093
View Documents
3M™
70071401874
Urethane Coated Belt Assembly, machine operation N/A
Only null left
29-635-2323
View Documents
3M™
70070515450
Comfort Belt Assembly, facility maintenance, machine operation, painting, sanding N/A
Only null left
Description
Description
  • For use with certain 3M respiratory headgear, filter/cartridges, batteries and breathing tubes
  • NIOSH approved respiratory system
  • Protects against certain gases, organic vapors, and particulates
  • Includes the components needed to make a complete 3M PAPR System when combined with 3M headgear and breathing tube:
    • Motor Blower
    • Flow Indicator and Adaptor
    • Blower Plugs
    • Web Belt, Urethane Coated Belt OR Comfort Belt
    • Power Coard
    • Battery Pack
    • Battery Charger
  • Single replaceable cartridge system
  • Contours to the hip and body for comfort
Specifications
Specifications
8 hours
PAPR Assembly, No Headtop
Motor blower, battery, charger, flow meter, power cord, blower plugs
High Efficiency (HE)
PAPR respirator
Belt-mounted
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product.