missing translation for 'onlineSavingsMsg'
Learn More

Gum arabic, powder

CAS: 9000-01-5

Supplier:  Thermo Scientific Chemicals 258850010

 View more versions of this product

Catalog No. AC258850010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Beige-Yellow to White
Powder
Total ash: < 4.0%
1.3500g/mL
Solubility in water: soluble. Other solubilities: insoluble in alcohol
Passes Test (per Eur. Pharm.)
0.1% max.
Plastic bottle
Gum arabic
9000-01-5
MFCD00081264
Acacia
1 kg
Authentic
15.0% max. (1 g, 105°C)
1.35

Chemical Identifiers

9000-01-5
MFCD00081264
Acacia
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 232-519-5

RTECSNumber : CE5945000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only