missing translation for 'onlineSavingsMsg'
Learn More

Wells Lamont™ GuardTec4 EliteGrip™ Gloves with Nitrile Palm

Hand protection with dexterity and fit

$156.17 - $156.17

Specifications

Color Gray
Cut Resistance Rating ANSI A4
Material Nitrile
Product Type Cut-resistant Gloves
For Use With (Application) Good grip in wet and oily environment and provides protection from lacerations and superior resistance against cuts
View More Specs

Products 5
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
Catalog Number Mfr. No. Glove Cuff Color Size Price Quantity  
19148839
View Documents
Wells Lamont™
Y9288XS
Yellow X-Small
Dozen of 12 for $156.17
Case of 12 Dozen for $1,703.64
Only null left
 
19148840
View Documents
Wells Lamont™
Y9288S
Red Small
Dozen of 12 for $156.17
Case of 12 Dozen for $1,606.32
Only null left
 
19148842
View Documents
Wells Lamont™
Y9288L
Brown Large
Dozen of 12 for $156.17
Case of 12 Dozen for $1,606.32
Only null left
 
19148843
View Documents
Wells Lamont™
WLCY9288XL
Blue X-Large
Dozen of 12 for $156.17
Case of 12 Dozen for $1,703.64
Only null left
 
19148844
View Documents
Wells Lamont™
Y9288XXL
Black 2X-Large
Dozen of 12 for $156.17
Case of 12 Dozen for $1,606.32
Only null left
 
Description

Description

  • Feature highly cut-resistant fiber that provides protection against laceration
  • Sandy nitrile palm coating provides excellent grip in wet and oily environments
  • Dark-colored shell conceals dirt and grime
  • Color: Gray

Specifications

Specifications

Gray
Nitrile
Good grip in wet and oily environment and provides protection from lacerations and superior resistance against cuts
ANSI A4
Cut-resistant Gloves
Knit
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'