Learn More
Mopec Guardian Rack System
Collapsible morgue storage rack system
Supplier: Mopec IR26401
Description
- Easily to assemble and disassemble – only 8 bolts
- Heavy duty design – Holds up to 1,500 lbs in body weight and lasts for many years and many assemble/disassemble cycles
- Suitable for refrigerated trailers or other cooled structures
- 3, 4 and 5 tier designs
- Large, heavy-duty casters are easy to roll and lockable
- Designed for loading using a forklift
- All stainless steel construction
Chemical Identifiers
| Stainless Steel | |
| 26 in. |
Specifications
| Stainless Steel | |
| 4 | |
| 26 in. |