missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Guardian™ 3000 Hotplate and Stirrer
Guardian™ 3000 Hotplate and Stirrer is designed for economical heating and stirring in all laboratory settings. Select from three plate sizes to suit your application. All models can be used with the optional temperature probe for precise temperature control of the sample.
$1,290.48 - $1,447.90
Specifications
| Voltage | 115 V |
|---|---|
| Controller Type | Digital |
| Frequency | 50/60 Hz |
| Type | Stirring Hotplate |
| Max. Sample Load | 15 L |
| Catalog Number | Mfr. No. | Top Plate Material | Temperature (Metric) | Size | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Top Plate Material | Temperature (Metric) | Size | Price | Quantity & Availability | |||
|
01918396
|
OHAUS™
30680266 |
Ceramic-coated Stainless Steel | Ambient +5° to 380°C | 135 mm dia. |
Each for $1,290.48
|
|
|||
|
01918394
|
OHAUS™
30680268 |
Ceramic | Ambient +5° to 500°C | 4 x 4 in. |
Each for $1,290.48
|
|
|||
|
01918395
|
OHAUS™
30680267 |
Ceramic | Ambient +5° to 500°C | 7 x 7 in. |
Each for $1,447.90
|
|
|||
Description
- Applications: Boiling Liquids, Water/Oil Bath Preparation, Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
- Display: Backlit LCD Display for temperature and speed, green indicator lights when unit is heating and stirring, prominent hot top indicator light, probe in use icon, Single Point Calibration icon
- Operation: Stirring from 80 to 1600 rpm, ambient temperature range from +5°C to 500°C (ceramic top) or 380°C (ceramic-coated steel top), power cord (included), stirring models include PTFE-coated stir bar
- Communication: N/A
- Construction: Durable metal housing, PET front panel, control knobs, on/off switch for heater disconnect, built-in support rod holder, easy-to-clean ceramic or ceramic-coated stainless steel tops
- Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring, temperature probe connector for use with optional temperature probe
Specifications
| 115 V | |
| 50/60 Hz | |
| 15 L | |
| 1 | |
| 80 to 1600 rpm | |
| Custom Backlit LCD |
| Digital | |
| Stirring Hotplate | |
| US | |
| 15 L | |
| 2 Years | |
| 80 to 1600 rpm |