missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Guardian™ 2000 Stirrer

Catalog No. 01918393
Change view
Click to view available options
Size:
135 mm dia.
4 x 4 in.
7 x 7 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
01918393 135 mm dia.
01918391 4 x 4 in.
01918392 7 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01918393 Supplier OHAUS™ Supplier No. 30680263

Guardian™ 2000 Stirrer is ideal for education and other lab environments that require affordable, everyday reliable stirring up to 15 L. All three sizes offer simple knob control to easily adjust the stirring functions.

  • Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
  • Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
  • Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
  • Communication: N/A
  • Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
  • Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring

Specifications

Top Plate Material Ceramic-coated Stainless Steel
Voltage 115 V
Controller Type Analog
Model e-G21STRDS
Size 135 mm dia.
Temperature (Metric) 5°C to 40°C, 80% RH, Non-condensing
Frequency 60 Hz
Wattage 35 W
Weight (English) 5.5 lb.
Weight (Metric) 2.5 kg
Type Stirring Hotplate
Shape Round
Max. Sample Load 15 L
Plug Type US
No. of Positions 1
Max. Container Size 15 L
Stirring Range 200 to 1600 rpm
Warranty 2 Years
Dimensions (L x W x H) 264 x 163 x 101 mm (10.4 x 6.4 x 4 in.)
Display Analog
Speed 200 to 1600 rpm
Show More Show Less