missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Guardian™ 2000 Stirrer
Description
- Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
- Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
- Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
- Communication: N/A
- Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
- Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
Specifications
Specifications
| Top Plate Material | Ceramic-coated Stainless Steel |
| Voltage | 115 V |
| Controller Type | Analog |
| Model | e-G21ST07R |
| Size | 7 x 7 in. |
| Temperature (Metric) | 5°C to 40°C, 80% RH, Non-condensing |
| Frequency | 60 Hz |
| Wattage | 35 W |
| Weight (English) | 6.2 lb. |
| Weight (Metric) | 2.8 kg |
| Show More |