missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Guardian™ 2000 Hotplate and Stirrer
Guardian™ 2000 Hotplate and Stirrer is ideal for education and other lab environments that require affordable, everyday fast heating and reliable stirring up to 15 L. All three sizes offer simple knob control to easily adjust the heating and stirring functions.
Supplier: OHAUS™ 30680259
Description
- Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
- Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
- Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
- Communication: N/A
- Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
- Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
Chemical Identifiers
| 115 V | |
| 60 Hz | |
| 15 L | |
| 1 | |
| 200 to 1600 rpm | |
| Analog |
| Analog | |
| Stirring Hotplate | |
| US | |
| 15 L | |
| 2 Years | |
| 200 to 1600 rpm |
Specifications
| Ceramic | |
| Analog | |
| 4 x 4 in. | |
| 158°F to 932°F | |
| 535 W | |
| 2.5 kg | |
| Square | |
| US | |
| 15 L | |
| 2 Years | |
| Analog |
| 115 V | |
| e-G21HS04C | |
| 70°C to 500°C | |
| 60 Hz | |
| 5.5 lb. | |
| Stirring Hotplate | |
| 15 L | |
| 1 | |
| 200 to 1600 rpm | |
| 264 x 163 x 110 mm (10.4 x 6.4 x 4.3 in.) | |
| 200 to 1600 rpm |