missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Guardian™ 2000 Hotplate

Guardian™ 2000 Hotplate is ideal for education and other lab environments that require affordable, everyday fast heating to 15 L. All three sizes offer simple knob control to easily adjust the heating functions.

$771.14 - $1,047.85

Specifications

Voltage 100/120 V
Controller Type Analog
Frequency 50/60 Hz
Type Stirring Hotplate
Max. Sample Load 15 L
View More Specs

Products 3
Catalog Number Mfr. No. Temperature (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Temperature (Metric) Size Price Quantity & Availability  
01918387
View Documents Promotion Details
OHAUS™
30680260
50°C to 380°C 135 mm dia.
Each for $1,047.85
Only null left
 
01918385
View Documents Promotion Details
OHAUS™
30680262
70°C to 500°C 4 x 4 in.
Each for $771.14
Only null left
 
01918386
View Documents Promotion Details
OHAUS™
30680261
70°C to 500°C 7 x 7 in.
Each for $865.57
Only null left
 
Description

Description

  • Applications: Boiling Liquids, Basic Sample and Solution Preparation, Dissolving Buffers and Reagents, Preparing Media, Concentrating Samples
  • Display: Prominent hot top indicator light, green indicator lights when unit is heating and stirring
  • Operation: Simple knob control, stirring up to 1600 rpm, temperature up to 500°C (ceramic top) or 380°C (ceramic coated stainless steel top), power cord (included), stirring models include PTFE coated stir bar
  • Communication: N/A
  • Construction: Durable metal housing, PET front panel, control knobs, ON/OFF switch for heater disconnect, built in support rod holder, easy to clean resin, ceramic or ceramic coated stainless steel tops
  • Design Features: Hot top indicator light to protect user, green indicator lights illuminate when unit is heating and stirring
Specifications

Specifications

100/120 V
50/60 Hz
15 L
1
2 Years
Analog
Stirring Hotplate
US
15 L
Analog
Videos
SDS
Documents

Documents