missing translation for 'onlineSavingsMsg'
Learn More

Guanylurea Sulfate 98.0+%, TCI America™

Catalog No. D043325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D043325G 25 g
D0433500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D043325G Supplier TCI America Supplier No. D043325G

Chemical Identifiers

CAS 591-01-5
Molecular Formula C2H6N4O
MDL Number MFCD00035565
Synonym Dicyandiamidine Sulfate

Specifications

Melting Point 190°C
Color White
Quantity 25 g
Formula Weight 302.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanylurea Sulfate
EINECSNumber (2)-1778
TSCA Yes