missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Guanosine 5'-triphosphate, disodium salt hydrate, 90%, for biochemistry

Catalog No. AC226252500
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226252500 250 mg Glass bottle
AC226250500 50 mg Glass bottle
AC226250010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226252500 Supplier Thermo Scientific Chemicals Supplier No. 226252500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Guanosine 5'-triphosphate, disodium salt hydrate, 90%, C10H14N5Na2O14P3.xH2O, CAS Number-56001-37-7 | CAS: 56001-37-7 | C10H14N5Na2O14P3 | 567.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Chemical Identifiers

CAS 56001-37-7
Molecular Formula C10H14N5Na2O14P3
Molecular Weight (g/mol) 567.14
MDL Number MFCD03410297
InChI Key FIZIYLKEXVIRHJ-KHRSEZDTNA-L
Synonym guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
PubChem CID 131676145
IUPAC Name disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]2O)C(=O)N1

Specifications

Absorbance 1.12 to 1.18 (A250/A260), 0.63 to 0.69 (A280/A260)
Melting Point 180°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble
Spectrophotometry 88% min. (lambda max. 252nm,pH 7)
Formula Weight 567.14
Percent Purity 90%
Related Substances 4% max. (typically) Guanosine diphosphate (HPLC)
Grade Biochemistry
Physical Form Crystalline Powder
Chemical Name or Material Guanosine 5'-triphosphate, disodium salt hydrate
Show More Show Less
EINECSNumber 259-940-7

RUO – Research Use Only