Learn More
Thermo Scientific Chemicals Guanosine-5'-triphosphate disodium salt
Description
Guanosine 5?-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF). Guanosine 5?-Triphosphate, Disodium Salt is a phosphoryl donor in protein synthesis and in signal transduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF). Guanosine 5′-Triphosphate, Disodium Salt is a phosphoryl donor in protein synthesis and in signal transduction.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Beilstein | 4113439 |
| Solubility Information | Soluble in water. |
| Formula Weight | 567.15 |
| Physical Form | Powder |
| Chemical Name or Material | Guanosine-5'-triphosphate disodium salt |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 259-940-7 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only