missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-monophosphate, Free acid, ≥98%, MP Biomedicals™

Catalog No. p-4774159
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10190880 100 mg
ICN10190890 500 mg
ICN10190891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10190880 Supplier MP Biomedicals Inc Supplier No. 0210190880
Only null left

Chemical Identifiers

CAS 85-32-5
Molecular Formula C10H14N5O8P
Molecular Weight (g/mol) 363.223
InChI Key RQFCJASXJCIDSX-UUOKFMHZSA-N
Synonym 5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate
PubChem CID 6804
ChEBI CHEBI:17345
IUPAC Name [(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N

Specifications

Color White
Assay Percent Range ≥98%
Quantity 100 mg
Formula Weight 363.2
Physical Form Powder
Chemical Name or Material Guanosine-5'-monophosphate, FTIR
EINECSNumber 201-598-8