missing translation for 'onlineSavingsMsg'
Learn More

Guanine 98.0+%, TCI America™

Catalog No. p-7130175
Change view
Click to view available options
Quantity:
1 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01691G 1 g
G016925G 25 g
G0169250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G01691G Supplier TCI America Supplier No. G01691G
Only null left

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Quantity 1 g
Formula Weight 151.13
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Guanine
EINECSNumber (5)-1136&(9)-0328
RTECSNumber MF8260000
TSCA Yes