missing translation for 'onlineSavingsMsg'
Learn More

Guanine, 99+%

Catalog No. p-3736633
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120250100 10 g Glass bottle
AC120250250 25 g Glass bottle
AC120251000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC120250100 Supplier Thermo Scientific Chemicals Supplier No. 120250100
Only null left

CAS: 73-40-5 | C5H5N5O | 151.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
IUPAC Name 2-amino-3,7-dihydropurin-6-one
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Absorbance lambda max. 248nm, pH 1, 1.35 to 1.41 (A250/A260), 0.82 to 0.86 (A280/A260), 0.47 to 0.51 (A290/A260)
Melting Point 360°C
Color Cream to White
Ignition Residue 0.1% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 26, 449
Merck Index 15, 4599
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in alkali hydroxides,dilute acids,sparingly soluble in alcohol and ether
Formula Weight 151.13
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Guanine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-799-8

RUO – Research Use Only