missing translation for 'onlineSavingsMsg'
Learn More

Guanine, 98%, Spectrum™ Chemical

Catalog No. 18610762
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610762 5 g Amber Glass Bottle
18610761 25 g Amber Glass Bottle
18610760 100 g Amber Glass Bottle
18610759 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610762 Supplier Spectrum Chemical Mfg Cor Supplier No. G10295GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

CAS Max % 1
Melting Point >300°C
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Guanine

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.