missing translation for 'onlineSavingsMsg'
Learn More

Guanine, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor G10291KG

 View more versions of this product

Catalog No. 18610759

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

73-40-5
>300°C
C5H5N5O
1 kg
NC1=NC(=O)C2=C(N1)N=CN2
151.13
Reagent
1
Amber Glass Bottle
MFCD00071533
UYTPUPDQBNUYGX-UHFFFAOYSA-N
2-amino-6,7-dihydro-3H-purin-6-one
0.98

Chemical Identifiers

73-40-5
151.13
UYTPUPDQBNUYGX-UHFFFAOYSA-N
NC1=NC(=O)C2=C(N1)N=CN2
C5H5N5O
MFCD00071533
2-amino-6,7-dihydro-3H-purin-6-one
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.