missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate (White Crystalline Powder), Fisher BioReagents™

Catalog No. BP2211
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2211 1 kg Poly Bottle
BP221250 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2211 Supplier Fisher BioReagents Supplier No. BP221 1
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the isolation of intact RNA from cell extracts

A more powerful denaturing agent than Guanidine Hydrochloride, Guanidine Thiocyanate is suitable for the isolation of intact RNA from cell extracts.

Chemical Identifiers

CAS 593-84-0
Molecular Formula C2H6N4S
Synonym Guanidinium Isothiocyanate

Specifications

Absorbance 0.8 max. (70% in water at 25°C) at 280nm
Melting Point 118°C
Color White
pH 5
Assay Percent Range ≥99.0 %
Packaging Poly Bottle
Linear Formula H2NC(:NH)NH2·HSCN
Quantity 1 kg
Solubility Information Soluble in water
Vapor Pressure Negligible
Formula Weight 118.16
Percent Purity ≥99.0%
Physical Form Powder or Solid
Chemical Name or Material Guanidine Thiocyanate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Harmful by inhalation, in contact with skin and if swallowed. Contact with acids liberates toxic gas. Harmful to aquatic organisms, may cause long-term advers
Recommended Storage RT