missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate 99.0+%, TCI America™

Catalog No. p-7130199
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G023025G 25 g
G0230500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G023025G Supplier TCI America Supplier No. G023025G
Only null left

Chemical Identifiers

CAS 593-84-0
Molecular Formula CH5N3
MDL Number MFCD00013027
Synonym Guanidinium Thiocyanate

Specifications

Melting Point 120°C
Color White
pH 4.5 to 7.0 (6M H2O 25°C)
Quantity 25 g
Formula Weight 118.16
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Thiocyanate
RTECSNumber XL1225000
TSCA Yes