missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Thiocyanate 98.0+%, TCI America™
Supplier: TCI America G036025G
Specifications
| Guanidine Thiocyanate [for Biochemical Research] | |
| 120°C | |
| 4.5 to 7.0 (6M H2O 25°C) | |
| MFCD00013027 | |
| 118.16 | |
| Crystalline Powder |
| 593-84-0 | |
| White | |
| CH5N3 | |
| 25 g | |
| ≥98.0% (T) |
Chemical Identifiers
| 593-84-0 | |
| CH5N3 | |
| MFCD00013027 |
Safety and Handling
RTECSNumber : XL1225000
TSCA : Yes