missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Sulfate 99.0+%, TCI America™

$63.38 - $469.75

Chemical Identifiers

CAS 594-14-9
Molecular Formula CH5N3
MDL Number MFCD00013115
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G016625G
View Documents
TCI America
G016625G
25 g
Each for $63.38
Only null left
 
G0166500G
View Documents
TCI America
G0166500G
500 g
Each for $469.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

594-14-9
CH5N3
MFCD00013115

Specifications

594-14-9
CH5N3
25 g
≥99.0% (T)
Guanidine Sulfate
White
MFCD00013115
216.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1773

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'