missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Sulfamate 90.0+%, TCI America™

Catalog No. p-7130104
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G002225G 25 g
G0022500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G002225G Supplier TCI America Supplier No. G002225G
Only null left

Chemical Identifiers

CAS 50979-18-5
Molecular Formula CH8N4O3S
Molecular Weight (g/mol) 156.16
MDL Number MFCD00060261
InChI Key LNEUSAPFBRDCPM-UHFFFAOYSA-N
PubChem CID 170947
IUPAC Name guanidine; sulfamic acid
SMILES NC(N)=N.NS(O)(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 156.16
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Sulfamate