missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Sulfamate 90.0+%, TCI America™

Catalog No. g002225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
G002225G 25 g
G0022500G 500 g
2 options

Catalog No. G002225G

Supplier: TCI America G002225G

Only null left

Chemical Identifiers

CAS 50979-18-5
Molecular Formula CH8N4O3S
Molecular Weight (g/mol) 156.16
MDL Number MFCD00060261
InChI Key LNEUSAPFBRDCPM-UHFFFAOYSA-N
PubChem CID 170947
IUPAC Name guanidine; sulfamic acid
SMILES NC(N)=N.NS(O)(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 156.16
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Sulfamate