missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydroiodide 97.0+%, TCI America™

Catalog No. G04501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04501G 1 g
G04505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04501G Supplier TCI America Supplier No. G04501G
Only null left

Chemical Identifiers

CAS 19227-70-4
Molecular Formula CH6IN3
Molecular Weight (g/mol) 186.984
InChI Key UUDRLGYROXTISK-UHFFFAOYSA-N
Synonym GAI, Guanidinium Iodide
PubChem CID 19820467
IUPAC Name guanidine;hydroiodide
SMILES C(=N)(N)N.I

Specifications

Melting Point 196°C
Color White
Quantity 1 g
Formula Weight 186.98
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Hydroiodide