missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Carbonate 98.0+%, TCI America™

Catalog No. G016125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G016125G 25 g
G0161500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G016125G Supplier TCI America Supplier No. G016125G

Chemical Identifiers

CAS 593-85-1
Molecular Formula CH5N3
MDL Number MFCD00013029

Specifications

Melting Point 333°C
Color White
Quantity 25 g
Formula Weight 180.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Carbonate
RTECSNumber FG1750000
TSCA Yes