Learn More
Guanidine carbonate, 98%
CAS: 593-85-1 | C3H12N6O3
$58.97 - $547.25
Chemical Identifiers
| CAS | 593-85-1 |
|---|---|
| Molecular Formula | C3H12N6O3 |
| MDL Number | MFCD00013029 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1285330
|
Thermo Scientific Chemicals
A1285330 |
250 g |
Each for $58.97
|
|
|||||
|
AAA128530B
|
Thermo Scientific Chemicals
A128530B |
1000 g |
Each for $157.64
|
|
|||||
|
AAA128530I
|
Thermo Scientific Chemicals
A128530I |
5000 g |
Each for $547.25
|
|
|||||
Description
It is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a strong organic alkali, organic intermediate, in soap and cosmetic products, and in the textile industry. It is also used to make fine chemicals. It is used in various industries to solve different purpose such as detergents as a synergistic agent, pharmaceuticals as an antibiotic and microencapsulation process for carbonless paper.
Solubility
Soluble in water 450 g/L (20°C); slightly soluble in alcohol and acetone.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 593-85-1 | |
| C3H12N6O3 | |
| MFCD00013029 |
Specifications
| 593-85-1 | |
| C3H12N6O3 | |
| 250 g | |
| Soluble in water 450g/L (20°C); slightly soluble in alcohol and acetone. | |
| 98% |
| ∼198°C (decomposition) | |
| MFCD00013029 | |
| 3628359 | |
| 180.17 | |
| Guanidine carbonate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 209-813-7
RTECSNumber : FG1750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only