missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guaiacol Carbonate 98.0+%, TCI America™
$57.79 - $1094.21
Chemical Identifiers
| CAS | 553-17-3 |
|---|---|
| Molecular Formula | C15H14O5 |
| Molecular Weight (g/mol) | 274.272 |
| MDL Number | MFCD00025723 |
| InChI Key | ORUJFMPWKPVXLZ-UHFFFAOYSA-N |
| Synonym | guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29 |
| PubChem CID | 11104 |
| IUPAC Name | bis(2-methoxyphenyl) carbonate |
| SMILES | COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC |
Chemical Identifiers
| 553-17-3 | |
| 274.272 | |
| ORUJFMPWKPVXLZ-UHFFFAOYSA-N | |
| 11104 | |
| COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC |
| C15H14O5 | |
| MFCD00025723 | |
| guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29 | |
| bis(2-methoxyphenyl) carbonate |
Specifications
| 553-17-3 | |
| White | |
| MFCD00025723 | |
| guaiacol carbonate, bis 2-methoxyphenyl carbonate, duotal, tussophob, guaiacyl carbonate, di-o-methoxyphenyl carbonate, bis-o-methoxyphenyl carbonate, carbonic acid bis 2-methoxyphenyl ester, guaiacol carbonic acid neutral ester, unii-q71xpq6r29 | |
| COC1=CC=CC=C1OC(=O)OC2=CC=CC=C2OC | |
| 274.272 | |
| 274.27 | |
| Crystalline Powder |
| 88°C | |
| C15H14O5 | |
| 25 g | |
| ORUJFMPWKPVXLZ-UHFFFAOYSA-N | |
| bis(2-methoxyphenyl) carbonate | |
| 11104 | |
| ≥98.0% (HPLC) | |
| Guaiacol Carbonate |
Safety and Handling
EINECSNumber : (9)-0330
TSCA : No