Learn More
Vector Laboratories GSL I B4, DyLight 594, 0.5 mg SA-5004-1
Catalog No. NC1795467
Encompass
Griffonia Simplicifolia Lectin I (GSL I) isolectin B4, DyLight 594, 0.5 mg, contains only the B subunits. It is a useful marker for endothelial cells from nonprimates, such as mouse, rat, rabbit, and goat, as well as a marker for non-peptidergic unmyelinated primary afferent neurons. This "B"-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues. DyLight 594 GSL I-B4 isolectin is produced by using special conjugation procedures to incorporate DyLight 594 into the affinity-purified lectin. This conjugate has an appropriate number of fluorochromes bound to provide the maximum fluorescence and optimum staining characteristics for this particular lectin. This reagent is supplied essentially free of unconjugated fluorochromes and inactive lectin. Excitation maximum: 592 nm; Emission maximum: is at 617 nm; Color: Red.
Catalog No. NC1795467 Supplier Vector Laboratories Supplier No. SA50041
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Griffonia Simplicifolia Lectin I (GSL I) isolectin B4, DyLight 594, 0.5 mg, contains only the B subunits. It is a useful marker for endothelial cells from nonprimates, such as mouse, rat, rabbit, and goat, as well as a marker for non-peptidergic unmyelinated primary afferent neurons. This "B"-rich lectin preferentially agglutinates blood group B cells and is specific for alpha-galactose residues. DyLight 594 GSL I-B4 isolectin is produced by using special conjugation procedures to incorporate DyLight 594 into the affinity-purified lectin. This conjugate has an appropriate number of fluorochromes bound to provide the maximum fluorescence and optimum staining characteristics for this particular lectin. This reagent is supplied essentially free of unconjugated fluorochromes and inactive lectin. Excitation maximum: 592 nm; Emission maximum: is at 617 nm; Color: Red.