missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals GSK-3 Inhibitor X

Catalog No. AAJ64659MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64659MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64659MCR Supplier Thermo Scientific Chemicals Supplier No. J64659MCR

For cellular and molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Common Name BIO-Acetoxime
Content And Storage 2°C to 8°C
Format Solid
Molecular Weight (g/mol) 398.21
Product Type GSK-3 Inhibitor X
Quantity 1 mg
Regulatory Status RUO
Sensitivity Light sensitive
Sequence (2'Z,3'E)-6-Bromoindirubin-3'-acetoxime

RUO – Research Use Only