Learn More
GSC Go Science Crazy Support Ring Clamp
Catalog No. S13755
Change view
Click to view available options
Diameter (English):
2 in. (Outer Dia.)
3 in. (Outer Dia.)
4 in. (Outer Dia.)
5 in. (Outer Dia.)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
S13755 4 in. (Outer Dia.)
S13753 2 in. (Outer Dia.)
S13754 3 in. (Outer Dia.)
S13756 5 in. (Outer Dia.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13755 Supplier GSC Go Science Crazy Supplier No. 4SRC4710
Not a toy. Product intended for ages 13 and up.
Great for holding all sorts of lab equipment in a small space. Works best with GSC Support Stands and Rods. The GSC Support Ring Clamp is a cast-iron clamp with nickel plated steel set screws. Can be clamped to a support stand rod for holding lab equipment.
  • Clamp made from cast iron
  • Nickel-plated steel screws
  • Great for support stands/rods
Specifications
Diameter (English) 4 in. (Outer Dia.)