missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSC Go Science Crazy Lead Cord with Banana and Alligator Plugs
Description
- With banana/alligator plugs on both ends
- For use with any electrical experiment
Specifications
Specifications
| Product Type | Banana/alligator |
| Width (English) | 24 in. |
| Color | Black |