Learn More
GSC Go Science Crazy Wimhurst Machine Economy
Catalog No. S43212A
Encompass
Change view
Click to view available options
Wimhurst machine
Catalog No.
S43212A Wimhurst machine
1 options
Catalog No. S43212A Supplier GSC Go Science Crazy Supplier No. 4509
Classic generator for producing electric charges.
  • Stand, Leyden jars, pulleys, and two handles are molded from durable plastic
  • Unit produces sparks approximately 2.5 in. long with appropriate weather conditions
  • Size: 14 x 14.5 x 8.5 in., weight: 6 lb.
Specifications
For Use With (Application) Classroom demonstrations by creating static electrical charge and high voltage sparks approximately 2.5 in. long with appropriate weather conditions
Weight (English) 6 lb.
Weight (Metric) 2.72 kg