Learn More
GSC Go Science Crazy Sintered Glass Funnels
Quality borosilicate glass funnel with porous filter plate.
Supplier: GSC Go Science Crazy FNLSINT802
Description
- Quality borosilicate glass funnel allows for fine filtration without the use of filter paper
Includes porous filter plate that is removable for cleaning and reuse
Chemical Identifiers
| Borosilicate Glass |
Specifications
| 80 mL | |
| Borosilicate Glass | |
| 2, Medium |