Learn More
GSC Go Science Crazy Happy and Sad Balls
A great demonstration that easily compares elastic and inelastic collisions. The GSC Happy and Sad Balls can be used to demonstrate the difference between elastic and inelastic collisions. These have virtually the same density, mass and appearance. When the balls are thrown down, one bounces while the other hits the surface and lies there.
Supplier:  GSC Go Science Crazy HSBL01
Catalog No. S13636
Only null left
Alert:
Not a toy. Product intended for ages 13 and up. May Contain Latex!
Description
Description
  • 2 balls of different materials
  • Compare elastic vs. inelastic
Specifications
Specifications
Neoprene Nosorex
Product Suggestions
Product Suggestions