Learn More
GSC Go Science Crazy Clay Triangle
The perfect tool for suspending a crucible. The GSC Clay Triangles are made for heating crucibles. These clay triangles are made with galvanized iron wires to help prevent premature rusting.
Supplier: GSC Go Science Crazy 4203012
Alert:
Not a toy. Product intended for ages 13 and up.
Description
- Great for heating crucibles
- Made with galvanized iron wires
- Rust resistant
- Note: The length shown in the specifications is the length of the sides
Specifications
| 2.5 in. | |
| 6.4 cm |