Learn More
GSC Go Science Crazy Clay Triangle
Description
- Great for heating crucibles
- Made with galvanized iron wires
- Rust resistant
- Note: The length shown in the specifications is the length of the sides
Specifications
Specifications
| Length (English) | 1.5 in. |
| Length (Metric) | 3.8 cm |