Learn More
GSC Go Science Crazy Clamp Holder
A cast iron clamp which can rotate, and is used to hold objects on a support rod
Supplier:  GSC Go Science Crazy 4CH39
 View more versions of this product
Catalog No. S13738
Only null left
Explore available promotions
Alert:
Not a toy. Product intended for ages 13 and up.
Description
Description
  • Cast iron clamp
  • Nickel-plated thumb screws
Specifications
Specifications
Right Angle Clamp
For use with GSC Support Stands and Rods
5/8 in.
Cast Iron, Nickel Plated (Screws)
Accept rods up to 0.625 in. dia.
Product Suggestions
Product Suggestions