Learn More
GSC Go Science Crazy Cast Iron Support Ring Stand
Great for holding all sorts of lab equipment in a small space. The GSC Cast Iron Support Ring Stand is made from cast-iron, with an enamel finish that is acid resistant. Includes a zinc plated steel rod that screws into the base support.
Supplier:  GSC Go Science Crazy 4SSC14
 View more versions of this product
Catalog No. S13745
Only null left
Alert:
Not a toy. Product intended for ages 13 and up.
Includes: Zinc plated steel rod that screws into the base support
Description
Description
  • Stand made from cast iron
  • Acid resistant
  • Plated steel support rod
  • Rod threaded to fit base
Specifications
Chemical Identifiers
Stand
Cast Iron
Specifications
Stand
5/16 in.
6 in.
Cast Iron
18 in.
4 in.
Product Suggestions
Product Suggestions