Learn More
GSC Go Science Crazy Alcohol Burner with Dual Wicks
Catalog No. S13728
Change view
Click to view available options
Includes:
Dual Wicks
Catalog No. Includes
S13728 Dual Wicks
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13728 Supplier GSC Go Science Crazy Supplier No. EPD42002
Edge
Not a toy. Product intended for ages 13 and up.
A great addition to any lab in which a small burner is needed. Made of durable metal to prevent breakage. The GSC Alcohol Burner with Dual Wicks is an all metal, permanent wick alcohol burner with a capacity 100ml tank. The wick portion is a ceramic fiber inserted into a thin copper tube. This construction allows for constant flame. The burner is fueled by high quality alcohol which can be found in most local hardware stores.
  • All metal to prevent breakage
  • Holds 100ml of alcohol
  • Constant flame construction
  • Ceramic fiber wick
Specifications
Product Type Alcohol Burner
Includes Dual Wicks